Learn More
Thermo Scientific™ Temperature Flow and Pressure Control Accessories
Product Code. 15560163 Shop All Thermo Scientific Products
Click to view available options
Type:
Flow Control Kit
Pressure Reducer
For Use With (Equipment):
ThermoFlex 7500 - 24000
ThermoFlex 900 - 10000
Size:
1 in. NPT
1/2 in. NPT
Unit Size:
Each
Product Code. 15560163
Brand: Thermo Scientific™ 611000000106
Only null left
Add to basket
Add to basket
Refine the pressure and flow characteristics of your Thermo Scientific™ recirculating chiller.
Customize the flow and pressure output of recirculating chiller to match the needs of your process. Available in several popular sizes Thermo Scientific™ Flow and pressure control kits for Thermo Scientific chillers can have you up and running without the hassle of finding parts.
  • Designed to work with your Thermo Scientific recirculating chiller
  • Available in popular sizes that match your recirculating chiller connections and interface with your process
  • Take the guess work out of finding parts to match your recirculating chiller connections
  • Designed to match the operating range of your recirculating chiller
Additional questions? Please contact Thermo Scientific Technical support if you’re unsure how best to refine the flow and pressure of your recirculating chiller.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ThermoFlex 900 - 10000
Type Flow Control Kit
Size 1/2 in. NPT