missing translation for 'onlineSavingsMsg'
Learn More

Hellma Flow-Through Cell, 137-QS

Highly pure Suprasil™ quartz glass flow-through cuvette for fluorescence analysis and designed with inlet/outlet tubes.

Brand:  Hellma 137-1-40

Product Code. 15888966

  • 487.88 € / Each
Estimated Shipment: 24-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High-purity synthetic fused silica quartz construction
  • UV/VIS range
  • Integrated inlet/outlet tubes
Specifications

Specifications

260 μL
9 mm
1/Pk
200 to 2500 nm
12.5 mm
Suprasil Quartz
Inlet/Outlet Tubes
1 mm
45 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hellma Flow-Through Cell, 137-QS > Path length: 1 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.