Learn More
Hellma Flow-Through Cell, 3-in-1, 177-QS
Highly pure Suprasil™ quartz glass flow-through cuvette for absorbance, transmission, and fluorescence measurements and designed with 2 FEP tubing screw connections.
Brand:  Hellma 176-765-85-40
 View more versions of this product
Product Code. 15786962
  • 1302.47 € / Each
Estimated Shipment: 22-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High-purity synthetic fused silica quartz construction
  • UV/VIS range
  • Integrated screw connections for FEP tubing
Specifications
Specifications
110 μL
6 mm
1/Pk
200 - 2500 mm
12.5 mm
Suprasil Quartz
Two Screw Connections, FEB Tubing
10 x 1 mm
35 mm