Learn More
Cole-Parmer Flow Through Tubing Weight, PTFE
Prevent tubing from coiling or floating during fluid transfer applications
86.25 €
Products 2
Product Code Brand Tubing Size Price Quantity & Availability  
Product Code Brand Tubing Size Price Quantity & Availability  
30053089
View Documents
Cole-Parmer
50117-98
3/16 in. to 1/4 in.
86.25 €
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
30054041
View Documents
Cole-Parmer
50117-99
5/16 in. to 7/16 in.
86.25 €
Each
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
Description
Description
Tapered hose barb fitting easily inserts into the inner diameter (ID) of flexible tubing. Keep tubing submerged in liquid, preventing floatation, and ensuring uninterrupted fluid flow. Constructed of chemically inert PTFE. Autoclavable—allowing for easy sterilization and reuse
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Cole-Parmer Flow Through Tubing Weight, PTFE