Learn More
Floxuridine, MedChemExpress
MedChemExpress Floxuridine (5-Fluorouracil 2'-deoxyriboside) is a pyrimidine analog and known as an oncology antimetabolite. Floxuridine inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating the ATM and ATR checkpoint signaling pathways in vitro. Floxuridine is a extreamly potent inhibitor for S. aureus infection and induces cell apoptosis. Floxuridine has antiviral effects against HSV and CMV.

99.90 € - 334.80 €
Specifications
Chemical Name or Material | Floxuridine |
---|---|
CAS | 50-91-9 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C9H11FN2O5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16472006
![]() |
Medchem Express
HY-B0097 |
500 mg |
334.80 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16448086
![]() |
Medchem Express
HY-B0097 |
100 mg |
99.90 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16411376
![]() |
Medchem Express
HY-B0097 |
200 mg |
151.20 €
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Floxuridine (5-Fluorouracil 2'-deoxyriboside) is a pyrimidine analog and known as an oncology antimetabolite. Floxuridine inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating the ATM and ATR checkpoint signaling pathways in vitro. Floxuridine is a extreamly potent inhibitor for S. aureus infection and induces cell apoptosis. Floxuridine has antiviral effects against HSV and CMV.Specifications
Floxuridine | |
White | |
C9H11FN2O5 | |
DMSO : 250 mg/mL (1015.48 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (203.10 mM) | |
246.19 | |
98.0% | |
Solid |
50-91-9 | |
Research | |
5-Fluorouracil 2'-deoxyriboside | |
OC[C@@H]1[C@H](C[C@H](N2C(NC(C(F)=C2)=O)=O)O1)O | |
246.19 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
H301
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.