missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Fluid Point Sampler

Designed to allow rapid point sampling from drums, kegs and most containers. Sampling Systems™ Fluid Point Sampler is ideal for sampling pharmaceuticals, cosmetics and food stuffs.

Brand:  Sampling Systems™ 5551S-1500

 View more versions of this product

Product Code. 15829371

  • 590.00 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured in 316L stainless steel with a silicone seal (FDA grade)
  • Available in a variety of lengths
  • Easy to clean
Specifications

Specifications

Stainless Steel (316L)
450 mL
1500 mm
Silicone Seal
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.