Learn More
Thermo Scientific™ Fluid Transfer Assembly, Adaptor Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Brand: Thermo Scientific™ SH31122.01
This item is not returnable.
View return policy
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
3.22 mm (1/8 in.) ID x 6.35 mm (1/4 in.) OD to 9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD; TPE 374; 2 m (6.6 ft); Luer to MPC | |
Adapter Set |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Thermo Scientific™ Fluid Transfer Assembly, Adaptor Set > Adaptor Set; 3.22mm (1/8 in.) ID x 6.35mm (1/4 in.) OD to 9.52mm (3/8 in.) ID x 15.88mm (5/8 in.) OD; TPE 374; 2 m (78.7 in.); luer to MPC
Spot an opportunity for improvement?Share a Content Correction