missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fluo-4 Calcium Imaging Kit

Fluo-4 Calcium Imaging Kit

Brand:  Molecular Probes™ F10489

Product Code. 15445300

  • 680.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Fluo-4 Calcium Imaging Kit has been designed for the specific detection of calcium flux by imaging applications. Upon binding of calcium, a large increase (>100-fold) in fluorescence emission can be detected. To minimize time to experimental results, this kit contains all of the reagents necessary for the detection of calcium flux from live cells.

Features of the Fluo-4 Calcium Imaging Kit include:

  • Emission from the calcium-bound Fluo-4 dye (ex/em 494/506 nm) can be detected with standard FITC filters
  • Upon binding calcium, fluorescence intensity increases >100-fold
  • Kd for Ca2+ in buffer: ∼335 nM
  • Contains all necessary reagents, reducing time to experimental results
  • Exhibits fluorescence increase upon binding calcium with little shift in wavelength

    All Necessary Reagents Included
    The Fluo-4 Calcium Imaging Kit contains all of the necessary components for the detection of calcium flux by imaging applications. In addition to Fluo-4 dye, the kit contains PowerLoad™ Concentrate, a 10X Neuro Background Suppressor solution, and probenecid.

    Labeled calcium indicators, such as Fluo-4 dye, are molecules that exhibit an increase in fluorescence upon binding calcium. Fluo-4 is an analog of Fluo-3, with two key chemical differences that result in an increased fluorescence excitation at 488 nm and consequently higher fluorescence signal levels (>100-fold increase in fluorescence intensity upon binding calcium). For ease of use, the Fluo-4 dye is supplied in a ready-to-use solution.

    For easy cell loading, the Fluo-4 Calcium Imaging Kit includes PowerLoad™ Concentrate. Due to the unique nature of the PowerLoad™ solution, it can be used in the prescence of complete culture media, thus reducing the negative effects of replacing media or loading in serum-free media.

    Baseline autofluorescence caused by components within growth media can be greatly reduced by the addition of the included Neuro Background Suppressor. This solution has been specifically formulated for use with neuronal cells and will not cause osmotic shock. Additionally, it has been used successfully with many different cell types to efficiently suppress background fluorescence without sacrificing the specific cellular fluorescence generated in the assay.

    A proprietary, water-soluble probenecid (which is commonly used to inhibit cellular transport and thus reduces the baseline signal) is also supplied with the Fluo-4 Calcium Imaging Kit.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Contains:
  • 50 μL of Fluo-4 AM Dye in DMSO
  • 500 μL of PowerLoad™ Concentrate
  • 5 mL of Neuro Background Suppressor
  • 1 vial of Probenecid
  • Store at 2°C to 8°C
  • Do not freeze
  • Calcium Imaging Kit
    Molecular Probes™
    Calcium Assay
    Calcium Indicator
    1 kit
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.