missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fluo-4 Direct™ Calcium Assay Kit

Fluo-4 Direct™ Calcium Assay Kit

Brand:  Molecular Probes™ F10473

3435.20 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10637215

  • 4520.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Fluo-4 Direct™ Calcium Assay Kit was formulated to provide a homogeneous fluorescent calcium assay that:
1. Easily loads into cells
2. Achieves a large assay window and
3. Suppresses background fluorescence generated from the calcium indicator in complete media with little-to-no impact on the specific cellular fluorescence

This advanced formulation allows the assay to be run in a simple addition only format in the presence of serum-containing media. Fluo-4 Direct™ is the third addition to the Fluo-4 family of calcium detection reagents. Fluo-4 AM and Fluo-4 NW both require media removal before assay detection, but Fluo-4 NW adds convenience by including the PowerLoad™ reagent for ease in cell loading. The Fluo-4 Direct™ Calcium Assay Kit is similar to Fluo-4 NW in that it is formulated with PowerLoad™ for ease in cell loading, but unique in that it can be used in the presence of complete culture media and will efficiently suppress background fluorescence without sacrificing the specific cellular fluorescence generated in the assay.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green
Fluorescence
Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
Fluorescent Dye-Based
Powder
Molecular Probes™
Room Temperature
  • 1 L Fluo-4 Direct™ Calcium Assay Reagent (Component A)
  • 1.54 g Probenecid (Component B)
  • Store at ≤-20°C. Dessicate and protect from light.
Cell Viability and Proliferation
Calcium Assay Kit
Visible
Chemical
1 x 1 L
Nucleus, Organelles, Cytoplasm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.