Learn More
Molecular Probes™ Fluo-4 NW Calcium Assay Kit
Fluo-4 NW Calcium Assay Kit
Brand:  Molecular Probes™ F36205
 View more versions of this product
Product Code. 10135132
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
*The Fluo-4 NW (no-wash) Calcium Assay Kits provide a simplified method for detecting intracellular calcium, without a quencher dye or a wash step.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store in freezer -5°C to -30°C and protect from light.
Calcium Assay
Stain
100 Microplate(s)
Fluorescence
Microplate Reader
Fluorescent Dye-Based
Room Temperature