Learn More
Thermo Scientific™ Fluophase™ PFP Javelin™ Guard Columns
Offer convenient, easy to use protection for analytical HPLC columns
Brand:  Thermo Scientific™ 82705-022106
 View more versions of this product
Product Code. 11553321
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Particle size: 5μm
Features of Fluophase PFP (Perfluorophenyl) Solid Phase
  • Perfluorinated phases provide unique selectivity
  • Extra retention and selectivity for halogenenated compounds
  • Excellent for taxane analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μm
20 mm
2.1 mm
PFP