missing translation for 'onlineSavingsMsg'
Learn More
Fluorene, 98+%, Thermo Scientific Chemicals Product Code.: 11301527

Fluorene, 98+%, Thermo Scientific Chemicals

Product Code. 11301527
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11301527

Brand: Thermo Scientific Alfa Aesar L02124.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes

Solubility
Soluble in hot alcohol, ether, benzene and glacial acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-73-7
Molecular Formula C13H10
Molecular Weight (g/mol) 166.22
MDL Number MFCD00001111
InChI Key NIHNNTQXNPWCJQ-UHFFFAOYSA-N
Synonym fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, floureneShow MoreShow Less
PubChem CID 6853
ChEBI CHEBI:28266
IUPAC Name 9H-fluorene
SMILES C1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 113°C to 117°C
Density 1.202
Boiling Point 295°C
Flash Point 151°C (303°F)
Quantity 100 g
UN Number UN3077
Beilstein 1363491
Merck Index 14,4155
Solubility Information Soluble in hot alcohol,ether,benzene and glacial acetic acid. Insoluble in water.
Formula Weight 166.22
Percent Purity ≥98%
Chemical Name or Material Fluorene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241100 Certificate TypeCertificate of Analysis Date10/06/2022 Catalog NumberL02124.14,L02124.36,L02124.22
Lot NumberF1787A Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL02124.14,L02124.36,L02124.22
Lot Number10236068 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02124.14,L02124.36,L02124.22
Lot Number10135353 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02124.14,L02124.36,L02124.22
Lot Number10207515 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02124.14,L02124.36,L02124.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fluorene, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.