Learn More
Fluorene, 98+%, Thermo Scientific Chemicals
Description
Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes
Solubility
Soluble in hot alcohol, ether, benzene and glacial acetic acid. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 113°C to 117°C |
Density | 1.202 |
Boiling Point | 295°C |
Flash Point | 151°C (303°F) |
Quantity | 500 g |
UN Number | UN3077 |
Beilstein | 1363491 |
Merck Index | 14,4155 |
Solubility Information | Soluble in hot alcohol,ether,benzene and glacial acetic acid. Insoluble in water. |
Formula Weight | 166.22 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10246250 | Certificate of Analysis | 09/05/2023 | L02124.36 |
10241100 | Certificate of Analysis | 10/06/2022 | L02124.14,L02124.36,L02124.22 |
F1787A | Certificate of Analysis | 08/12/2021 | L02124.14,L02124.36,L02124.22 |
10236068 | Certificate of Analysis | 07/12/2021 | L02124.14,L02124.36,L02124.22 |
10135353 | Certificate of Analysis | 07/12/2021 | L02124.14,L02124.36,L02124.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.