missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ FluoReporter™ Biotin Quantitation Assay Kit, for biotinylated proteins

FluoReporter™ Biotin Quantitation Assay Kit, for biotinylated proteins

Brand:  Molecular Probes™ F30751

189.42 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10462282

  • 287.00 € / 1 set
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The FluoReporter™ Biotin Quantitation Assay Kit provides a sensitive fluorometric assay for determining the number of biotin labels on nucleic acid samples (i.e., cDNA). The assay uses Biotective™ Green reagent, which contains a fluorescent dye that is quenched by ligands occupying the biotin binding sites. In the presence of biotin, the quencher dye ligands are displaced and fluorescence occurs. The fluorescence is proportional to the amount of added biotin. The results can be read with any fluorescence-based microplate reader capable of detecting FITC dye.

Important Features of FluoReporter™ Biotin Quantitation Assay Kits:
  • Can detect from 4 to 80 picomoles of biotin in a sample (50-fold higher sensitivity than HABA biotin-binding assay)
  • Can be applied to as little as 13 ng of biotin-labeled nucleic acid (more is needed for lower degrees of labeling)
  • Excitation/emission maxima of the reagent is 495/519 nm
  • Supplied with a biotinylated positive control for standard curve

    For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Biotin Quantitation Assay Kit
    Store in freezer (-5 to -30°C) and protect from light.
    Room Temperature
    Biotin
    1 Kit
    Fluorescence
    FluoReporter™
    Proteins
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.