Learn More
Fluorescamine, Thermo Scientific Chemicals
Description
For fluorescent detection of amino acids, peptides and proteins.Fluorescamine plays an important role as a reagent for the detection of amines, peptides and proteins. It is also used as a substrate for measuring protease activity. It is utilized in the detection and quantitation of residual aminopenicillins and sulfonamides in honey by HPLC. It finds application in the determination of lisinopril in human plasma and urine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 155°C to 158°C |
Color | White to Cream |
Quantity | 25 mg |
Merck Index | 14,4158 |
Solubility Information | Soluble in acetone,ethanol,chloroform,dimethylsulfoxide and acetonitrile. Slightly soluble in water. |
IUPAC Name | 4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione |
Formula Weight | 278.27 |
Physical Form | Powder |
Chemical Name or Material | Fluorescamine |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P05L009 | Certificate of Analysis | 07/03/2025 | 043749.MC,043749.MB,043749.ME |
10252731 | Certificate of Analysis | 10/10/2024 | 043749.MC,043749.MB,043749.ME |
U09K009 | Certificate of Analysis | 14/08/2024 | 043749.MC,043749.MB,043749.ME |
P20K013 | Certificate of Analysis | 02/04/2024 | 043749.MC,043749.MB,043749.ME |
10249972 | Certificate of Analysis | 12/03/2024 | 043749.MC,043749.MB,043749.ME |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.