Learn More
Cytiva Fluoro Rubber/NBR/Polyamide/Polyethylene/Polypropylene/Stainless Steel material AC Flow Adapter
Basic C columns are of fixed volume but can be upgraded by adding these flow adapters
Brand:  Cytiva 19-5207-01
Product Code. 10670171
  • 247.00 € / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The basic C columns are of fixed volume but can be upgraded by adding these flow adapters (which keep bed height constant and protect from insoluble particles) and a thermostatic jacket. Three adjustable flow adapters are available for the C column series (AC 10, AC 16, and AC 26), allowing the choice of bed height to suit the application. Flow adapters are available for each of the column diameters to allow optimal bed heights for each application, as are thermostatic jackets for precise temperature control.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
60°C
1 Pack
14.5 psi