missing translation for 'onlineSavingsMsg'
Learn More

Fluoroacetone, 97%, Thermo Scientific Chemicals

Product Code. 10661365
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 430-51-3
Molecular Formula C3H5FO
Molecular Weight (g/mol) 76.06
MDL Number MFCD00000451
InChI Key MSWVMWGCNZQPIA-UHFFFAOYSA-N
Synonym fluoroacetone, 1-fluoro-2-propanone, 1-fluoroacetone, 2-propanone, 1-fluoro, mono-fluoroacetone, acetone, fluoro, 1-fluoroacetone #, ch3coch2f, acmc-1bne0
PubChem CID 9889
IUPAC Name 1-fluoropropan-2-one
SMILES CC(=O)CF

Specifications

Color Red
Density 1.0540g/mL
Boiling Point 75.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH2F
Refractive Index 1.3660 to 1.3700
Quantity 5 g
Specific Gravity 1.054
Formula Weight 76.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Fluoroacetone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Fluoroacetone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'