missing translation for 'onlineSavingsMsg'
Learn More

Fluorobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11416987
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Fluorobenzene is used as a reagent for plastic or resin polymers. It acts as a solvent for highly reactive species. On fluorination of fluorobenzene it gives 1,2-difluorobenzene. It is also used as a material in making medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorobenzene is used as a reagent for plastic or resin polymers. It acts as a solvent for highly reactive species. On fluorination of fluorobenzene it gives 1,2-difluorobenzene. It is also used as a material in making medicine.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-06-6
Molecular Formula C6H5F
Molecular Weight (g/mol) 96.10
MDL Number MFCD00000280
InChI Key PYLWMHQQBFSUBP-UHFFFAOYSA-N
Synonym benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene
PubChem CID 10008
ChEBI CHEBI:5115
IUPAC Name fluorobenzene
SMILES FC1=CC=CC=C1

Specifications

Melting Point -42°C
Density 1.025
Boiling Point 84°C to 85°C
Flash Point −15°C (−5°F)
Odor Aromatic
Refractive Index 1.466
Quantity 250 g
UN Number UN2387
Beilstein 1236623
Merck Index 14,4170
Solubility Information Slightly soluble in water.
Formula Weight 96.1
Percent Purity 99%
Chemical Name or Material Fluorobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Fluorobenzene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Fluorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.