Learn More
Gibco™ FluoroBrite™ DMEM

Gibco™ FluoroBrite™ DMEM features a background fluorescence that is comparable to PBS and 90% lower than that emitted by standard phenol red–free DMEM.
46.14 € - 395.00 €
Specifications
Classification | Animal Origin-free |
---|---|
Concentration | 1 X |
Form | Liquid |
Product Type | DMEM (Dulbecco's Modified Eagle Medium) |
Serum Level | Standard Serum Supplementation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15291866
![]() |
Gibco™
A1896701 |
500 mL |
46.14 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15266695
![]() |
Gibco™
A1896702 |
10 x 500 mL |
395.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gibco™ FluoroBrite™ DMEM features a background fluorescence that is comparable to PBS and 90% lower than that emitted by standard phenol red–free DMEM. Formulated to include the required nutrients for routine cell culture when supplemented with 10% fetal bovine serum and 4 mM L-glutamine or GlutaMAX™ Supplement, FluoroBrite™ DMEM is designed to enhance the signal-to-noise ratio of fluorophores, enabling researchers to visualize even the weakest fluorescent events in an environment that promotes optimum cell health. Additional features include:
- Enhancement of fluorescence signal during live-cell imaging
- DMEM-based to help preserve cell health
Live-cell fluorescence microscopy is an essential technique for the visualization of fundamentally important and physiologically relevant biological events. A key challenge with this technique is the ability to image weak fluorophores without causing cell damage, photobleaching, or undesirable changes to cell health. FluoroBrite™ DMEM helps address these issues.
Specifications
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
High Glucose | |
cGMP-compliant under the ISO 13485 standard |
1 X | |
DMEM (Dulbecco's Modified Eagle Medium) | |
Sterile-filtered | |
No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate | |
Store in refrigerator (2–8°C). Protect from light. |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.