missing translation for 'onlineSavingsMsg'
Learn More

Fluorocyclohexane, 97%, Thermo Scientific Chemicals

Product Code. 15408979
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Fluorocyclohexane is used as an intermediate in chemical research and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorocyclohexane is used as an intermediate in chemical research and pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Store in cool, dry c
TRUSTED_SUSTAINABILITY

Specifications

CAS 372-46-3
Melting Point 13°C
Density 0.928
Boiling Point 103°C to 105°C
Flash Point 5°C (41°F)
Molecular Formula C6H11F
Refractive Index 1.416
MDL Number MFCD00021285
Quantity 5 g
UN Number UN1993
Beilstein 1900795
Synonym cyclohexane, fluoro, cyclohexyl fluoride, cyclohexylfluoride, fluoro-cyclohexane, acmc-1cpcv
Solubility Information Not miscible or difficult to mix in water.
InChI Key GOBGVVAHHOUMDK-UHFFFAOYSA-N
SMILES C1CCC(CC1)F
IUPAC Name fluorocyclohexane
Molecular Weight (g/mol) 102.152
PubChem CID 78988
Formula Weight 102.15
Percent Purity 97%
Chemical Name or Material Fluorocyclohexane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'