missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Flupirtine maleate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Flupirtine maleate is a non-opioid analgesic with muscle relaxant properties. Activates KV7 potassium channels, indirectly antagonizes NMDA receptors and modulates GABAA receptors. Exhibits neuroprotective actions in a model of cerebral ischemia in mice and reduces apoptosis and necrosis induced by noxious stimuli.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 420.4 |
| Purity | >98% |
| Molecular Weight (g/mol) | 420.397 |
| Chemical Name or Material | Flupirtine maleate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction