Learn More
FMOC isothiocyanate, 97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 44.0°C to 45.0°C |
Color | Yellow to Brown |
Elemental Analysis | 65.6 to 71.0 % (C) 4.8 to 5.2 % (N) |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Quantity | 5 g |
Formula Weight | 281.34 |
Percent Purity | 97% |
Physical Form | Liquid |
Chemical Name or Material | FMOC isothiocyanate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Safety and Handling


- FMOC isothiocyanate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.