Learn More
Cole-Parmer™ Foam-Tipped Short Glassware Brush
Foam-Tipped Short Glassware Brush - Excellent for all wide mouth bottles, culture, volumetric graduated, filtering and many other kinds of long flasks, jugs and cylinders. Brushes are tough, yet non-abrasive, and dishwasher safe. It works equally well on both plastic and glass bottles. Use with hot water. No detergents needed.
Brand:  Cole-Parmer™ 84552-14
19.98 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.
Product Code. 15798619
  • 19.60 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Ergonomically designed wide, flat handles - makes for easy rotating of brush
  • Polypropylene and foam bristles, wire and plastic handle
  • Non-abrasive
  • No detergents needed
  • Dishwasher safe
Specifications
Specifications
Polypropylene, Foam
Cleaning glass and plastic bottles, culture flasks, volumetric flasks, filter flasks etc., jugs and cylinders
2 in.
Wire, Plastic
13 in.