missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folin and Ciocalteu's Phenol Reagent, 2.0N, MP Biomedicals
Brand: MP Biomedicals 0219518690
169.30 EUR valid until 2024-12-31
Use promo code "21986" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Folin and Ciocalteu's Phenol Reagent | |
2.0N | |
10102-25-7, 10102-40-6, 7647-01-0, 10213-10-2, 7664-38-2 | |
500 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Folin and Ciocalteu's Phenol Reagent
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Lithium Sulfate Monohydrate, hydrochloric acid, Sodium Tungstate, Dihydrate, ACS, phosphoric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction