missing translation for 'onlineSavingsMsg'
Learn More

Folinic acid, calcium salt pentahydrate, 95.0-105.0%, Thermo Scientific Chemicals

Product Code. 10411081
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6035-45-6
Molecular Formula C20H21CaN7O7·5H2O
Molecular Weight (g/mol) 601.58
MDL Number MFCD00149465
InChI Key NPPBLUASYYNAIG-UHFFFAOYSA-L
Synonym folinic acid calcium pentahydrate
PubChem CID 131674093
IUPAC Name calcium;4-[[4-[(2-amino-5-formyl-4-oxido-7,8-dihydro-6H-pteridin-6-yl)methylamino]benzoyl]amino]-5-hydroxy-5-oxopentanoate;pentahydrate
SMILES C1C(N(C2=C(N1)N=C(N=C2[O-])N)C=O)CNC3=CC=C(C=C3)C(=O)NC(CCC(=O)[O-])C(=O)O.O.O.O.O.O.[Ca+2]

Specifications

Melting Point 240.0°C to 250.0°C
Color White-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 4249
Specific Rotation + 14.30
Solubility Information Solubility in water: freely soluble in water. Other solubilities: practically insoluble in alcohol
Formula Weight 601.58
Percent Purity 95 to 105%
Physical Form Powder
Chemical Name or Material Folinic acid, calcium salt pentahydrate, 95.0-105.0%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.