missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Food and Beverage Sign and Label Printer Kit for UK

Increase the safety and efficiency of your food or beverage plant with this complete safety sign and label printer kit.

Brand:  Brady™ 311115

Product Code. 17595146

  • 2530.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Easily design and print professional food or beverage safety signs, pipe markers, and labels when you need them
  • Print incredibly fast with automatic setup, built-in keyboard, and touchscreen interface
  • Create reliable labels that are washdown resistant and do not crumble when exposed to cleaning agents or pressure
  • Fully protect food processing with metal detectable signs that enable additional protection against contamination
Specifications

Specifications

LCD Touchscreen
12 in.
9.5 in.
10 in.
Not Autoclavable
Monochrome
256 MB
76 mm/s
Brady™ Workstation Basic Design Suite
9.1 kg
Component and Equipment Labeling, General Identification, Hazard Identification, Temperature Labeling
AC Adaptor
S3100-QY-UK-FnB
Thermal Transfer
305 mm
241 mm
260 mm
Non-sterile
Bulgarian, Chinese (Simplified), Chinese (Tradditional), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Korean, Norwegian, Polish, Portuguese, Russian, Romanian, Slovakian, Slovenian, Spanish, Swedish, Turkish
S3100
300 dpi
Ethernet, USB
UL, cUL, CE, WEEE, RoHS, RCM, EAC, CCC, RoHS 2
1 Year
110/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.