Learn More
Buerkle™ Forceps, Stainless Steel, Sharp, Bent Form
The Forceps, stainless steel, sharp, bent form with angled shape allow for accurate work in the laboratory. The angled laboratory tweezers are suitable for use in chemistry, the pharmaceutical industry, cosmetics and food & beverage.
Brand: Buerkle™ 5386-0102
4.66 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Stainless steel (1.4301)
- The pointed, fluted ends can be used to work reliably with sensitive materials, tissue or samples
- Flutings on the handle ensure a slip-free grip of the stainless steel tweezers
- Suitable for use in chemistry, the pharmaceutical industry, cosmetics and food & beverage
Specifications
Sharp, bent form | |
5.12 in. | |
130 mm | |
Forceps |
1 | |
Stainless Steel (1.4301) | |
Food |