Learn More
Manutan Fork extension, W x D 100 x 50mm, extended length 1830mm
Brand: Manutan A338743

Description
Safely lengthens forks for longer loads, held in place by integral retaining bar. Manufactured from heavy duty 8mm thick steel. Conforms to ISO 13284 British standards with test certificate supplied.
- Effective length (mm): 1830mm
- Maximum fork (W x D): 100mm x 50mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.