Learn More
Thermo Scientific™ Forma™ Inner Door Kits
Reduce cold air loss and improve temperature recovery after door closings when you choose factory-installed Thermo Scientific™ Forma™ Inner Door Kits for your ultra-low upright freezer.
Brand:  Thermo Scientific™ 1950547
 View more versions of this product
Product Code. 11376341
  • 674.36 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Reduce cold air loss and improve temperature recovery after door closings with Thermo Scientific™ Forma™ Inner Door Kits, a factory-installed door alternative option for your ultra-low upright freezer.
Four interior doors are already included on our -86°C uprights. The five inner door option for 8600 and 900 Series includes five powder-coated inner doors and one additional shelf.
Ordering Information:
Factory installed. Must be ordered with the refrigerator or freezer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ultra-Low Freezer Inner-Door
489.8 L
5
1
Forma™ 7000 and 900 Series, 17.3 cu. ft. Uprights
17.3 cu. ft.
-86°C
Powder-coated Steel
Upright