missing translation for 'onlineSavingsMsg'
Learn More

Formal Buffered Saline, Reagecon
BIOPHARMA

Fixative mixture of formaldehyde in isotonic saline for routine histopathology.

14.60 € - 56.40 €

Specifications

Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11503202
View Documents
Reagecon
FBS1
1 L HDPE Bottle GL45
14.60 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11593192
BIOPHARMA
View Documents
Reagecon
FBS5
5 L HDPE Jerrycan
56.40 €
5 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Fixative for histopathology.
Specifications

Specifications

Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • FORMAL BUFFERED SALINE

Signal Word
  • Danger

Hazard Category
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde, Sodiumdihydrogenphosphate

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.