missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Formal-Fixx™
Acquire amazing results with Epredia™ Formal-Fixx™, which is excellent for immunochemistry.
Brand: Epredia™ 6764255
60.20 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Enhances H&Es and special stains
- Minimizes the need to enzyme digest
- Reduces protein cross-linking
- Produces well-defined nuclear detail
Specifications
Fixative | |
3.8L concentrate (makes 19L total) |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H370-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : formaldehyde, zinc sulphate, methyl alcohol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction