missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol

Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant.

35.48 € - 344.72 €

Specifications

Chemical Name or Material Formaldehyde
Name Note 37 wt% Solution in Water, Stabilized with 10 to 15% Methanol
CAS 50-00-0, 7732-18-5, 67-56-1
Melting Point -15.0°C
Color Clear
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10426730
View Documents
Thermo Scientific Chemicals
119690010
1 L Plastic bottle
35.48 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10041040
View Documents
Thermo Scientific Chemicals
119690025
2.5 L Plastic bottle
69.75 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10182860
View Documents
Thermo Scientific Chemicals
119690050
5 L Plastic Jerrycan
96.58 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10638333
View Documents
Thermo Scientific Chemicals
119690250
25 L Plastic Jerrycan
344.72 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; that can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol

Applications

  • It May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.