missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol

Product Code. 10647021 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
5kg
This item is not returnable. View return policy

Product Code. 10647021

Brand: Thermo Scientific Chemicals 410730010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant. | CAS: 50-00-0 | CH2O | 30.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol

Applications

  • May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formaldehyde
Name Note 37 wt% Solution
CAS 50-00-0, 7732-18-5, 67-56-1
Melting Point -15.0°C
Color Clear
Density 1.0830g/mL
Boiling Point 97.0°C
Flash Point 50°C
Ignition Residue 0.005% max
Packaging Plastic Bottle
Molecular Formula CH2O
Linear Formula HCHO
Refractive Index 1.3755 to 1.3775
MDL Number MFCD00003274
Quantity 1 kg
Fieser 01,397; 02,200; 04,238; 05,312; 06,264; 07,158; 08,231; 09,224; 10,186; 11,240; 12,232; 14,168; 15,161; 16,171
Merck Index 15, 4263
Specific Gravity 1.083
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and acetone
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES C=O
IUPAC Name formaldehyde
Stabilizer 10 to 15% methanol
Molecular Weight (g/mol) 30.03
PubChem CID 712
ChEBI CHEBI:16842
Formula Weight 30.03
Vapor Pressure 2mbar at 20°C
Percent Purity 36.5 to 38.0%
Grade ACS Reagent
Titratable Acid 0.006 meq/g max
Physical Form Solution
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Formaldehyde, 37 wt% solution, stabilized with methanol
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Sicherheitshinweis
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Formaldehyde, Methylalcohol

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt