missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Pure, Solution, 10% (v/v) In 0.9% Sodium Chloride Solution, Fisher Chemical

Product Code. 10539190
Click to view available options
Quantity:
5 L
Packaging:
Plastic container
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Physical Form Liquid
Quantity 5 L
MDL Number 3274
Packaging Plastic container
compliance-icons compliance-icons
Product Identifier
  • Formaldehyde solution 10% v/v in 0.9% NaCl solution
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde, Methylalcohol, Sodium chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'