missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Solution, Contains 10-15% Methanol, Honeywell™
Contains 10-15% methanol as stabilizer, 37 wt. % in H2O
43.63 € - 188.97 €
Identificadores de productos químicos
CAS | 50-00-0 |
---|---|
Molecular Formula | CH2O |
Molecular Weight (g/mol) | 30.03 |
MDL Number | MFCD00003274 |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
Synonym | formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde |
PubChem CID | 712 |
ChEBI | CHEBI:16842 |
IUPAC Name | formaldehyde |
SMILES | C=O |
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
15618490
|
Honeywell Research Chemicals
F1635-25ML |
25 mL | Poly Bottle | N/A | |||||
15627860
|
Honeywell Research Chemicals
F1635-500ML |
500 mL | Poly Bottle | N/A | |||||
15628490
|
Honeywell Research Chemicals
F1635-4L |
4 L | Poly Bottle |
188.97 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Identificadores de productos químicos
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Especificaciones
50-00-0 | |
Colorless | |
3.5 to 4.5 (at 20°C) | |
64°C (147.2°F) | |
HCHO | |
UN2209 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
C=O | |
30.03 | |
CHEBI:16842 | |
Formaldehyde solution, 37 wt. % in H2O |
<-15°C | |
1.09g/mL at 25°C | |
93°C to 96°C (1013 hPa) | |
CH2O | |
MFCD00003274 | |
1209228 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
30.03g/mol |
Seguridad y manipulación
P201-P260-P280-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P308 + P311
missing translation for 'dotInformation' : 8/PGIII
Nombre del producto
Palabra de advertencia
Clasificación
Indicaciones de peligro
Consejos de prudencia
Otros peligros
- Formaldehyde solution
Palabra de advertencia
- Danger
Clasificación
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 3
Indicaciones de peligro
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H370-Causes damage to organs.
Consejos de prudencia
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Otros peligros
- MIXTURE LIST-Contains : formaldehyde, methanol