missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Solution, Honeywell
45.19 € - 604.19 €
Chemical Identifiers
CAS | 50-00-0 |
---|---|
Molecular Formula | CH2O |
Molecular Weight (g/mol) | 30.03 |
MDL Number | MFCD00003274 |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
Synonym | formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde |
PubChem CID | 712 |
ChEBI | CHEBI:16842 |
IUPAC Name | formaldehyde |
SMILES | C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15631470
|
Honeywell Riedel-de Haën
15513-1L |
1 L |
45.19 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15641470
|
Honeywell Riedel-de Haën
15513-2.5L |
2.5 L |
85.43 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15661470
|
Honeywell Riedel-de Haën
15513-60KG |
60 kg |
604.19 €
60kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Specifications
50-00-0 | |
64°C (147.2°F) | |
HCHO | |
2209 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
C=O | |
30.03 | |
CHEBI:16842 | |
Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP |
1.09g/mL at 25°C | |
CH2O | |
MFCD00003274 | |
1209228 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
30.03g/mol |
Safety and Handling
P201-P260-P280-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P308 + P311
H301 + H311 + H331-H314-H317-H335-H341-H350-H370
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Formaldehyde solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H370-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : formaldehyde, methanol