missing translation for 'onlineSavingsMsg'
Learn More

Formaldoxime trimer hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 11428910
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6286-29-9
Molecular Formula C3H10ClN3O3
Molecular Weight (g/mol) 171.58
MDL Number MFCD00012778
InChI Key VRHJXINSVWQXEB-UHFFFAOYSA-N
Synonym formaldoxime trimer hydrochloride, sym-triazinetriol hydrochloride, triformoxime hydrochloride, 1,3,5-triazinane-1,3,5-triol hydrochloride, 1,3,5-triazaperhydroine-1,3,5-triol, chloride, 1,3,5-triazine, hexahydro-1,3,5-trihydroxy-, hydrochloride
PubChem CID 15919123
IUPAC Name 1,3,5-trihydroxy-1,3,5-triazinane;hydrochloride
SMILES Cl.ON1CN(O)CN(O)C1

Specifications

Melting Point ∼128°C (decomposition)
Quantity 10 g
UN Number UN2811
Beilstein 3690722
Sensitivity Hygroscopic
Formula Weight 171.57
Percent Purity 95%
Chemical Name or Material Formaldoxime trimer hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.