missing translation for 'onlineSavingsMsg'
Learn More

Formamide, ≥99.0% (GC), Reagent Grade, Honeywell™

Product Code. 15648490
Click to view available options
Quantity:
5 mL
100 mL
250 mL
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99.0% (GC)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

pH 4.0 to 5.0 (at 20°C)
Melting Point 1°C to 3°C
Color Colorless
Density 1.134g/mL at 25°C
Boiling Point 210°C (1013 hPa)
Quantity 5 mL
Flash Point 175°C (347°F)
Ignition Point 932°F
Odor Weak
Packaging Glass Bottle
Physical Form Liquid
Linear Formula HCONH2
Vapor Pressure 0.08mmHg (20°C), 30mmHg (129°C)
UN Number NONH for all modes of transport
Beilstein 505995
Solubility Information Fully miscible in water
Formula Weight 45.04g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material Formamide
Show More Show Less
compliance-icons
Product Identifier
  • Formamide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'