missing translation for 'onlineSavingsMsg'
Learn More
Formamide, ACS, 99.5+%, Thermo Scientific Chemicals Product Code.: 11367088

Formamide, ACS, 99.5+%, Thermo Scientific Chemicals

Product Code. 11367088
4 x 1 kg
Click to view available options
Quantity:
250 g
1 kg
4 x 1 kg
This item is not returnable. View return policy

Product Code. 11367088

Brand: Thermo Scientific Alfa Aesar 014835.D6

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formamide is used for cryopreservation of tissues and organs and in hybridization buffers. It acts as an RNA stabilizer in gel electrophoresis by deionizing RNA. It is used as an alternative solvent for the electrostatic self-assembly of polymer nanofilms. It is used in the preparation of brain sections of mice for BrdU immunohistochemistry and fish analysis of HeLa cells. It essentially plays a vital role in crystallization of pharmaceuticals, in soil stabilization, as a solvent in inks, as a component of liquid fertilizers, as a heat-resistant coating and in some personal care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2°C
Color Colorless
Density 1.134 g/mL
Boiling Point 210°C
Quantity 4 x 1 kg
Flash Point 154°C (310°F)
Odor Ammonia-like
Physical Form Liquid
Refractive Index 1.4475
Assay Percent Range >99.5%
Linear Formula HCONH2
Beilstein 505995
Merck Index 14,4237
Solubility Information Miscible with water.
Formula Weight 45.04
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Formamide, For ACS analysis
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR16C704 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number014835.D6,014835.30,014835.A1
Lot NumberJ05U895 Certificate TypeCertificate of Analysis Date30/06/2022 Catalog Number014835.D6,014835.30,014835.A1
Lot NumberP30E712 Certificate TypeCertificate of Analysis Date26/05/2022 Catalog Number014835.D6,014835.30,014835.A1
Lot NumberZ28D754 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number014835.D6,014835.30,014835.A1
Lot NumberH18Y805 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number014835.D6,014835.30,014835.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formamide, ACS, 99.5+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.