missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Plastic Container
Plastic-coated Glass Bottle
Description
Formamide, Certified AR for Analysis

Specifications
Specifications
pH | 4 to 5 |
Melting Point | 2°C |
Boiling Point | 210°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 0.08mbar at 20°C |
Viscosity | 3.75 mPaS at 20°C |
Formula Weight | 45.04g/mol |
Safety and Handling

Product Identifier
- Formamide
- Danger
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- H351-Suspected of causing cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Formamide, Certified AR for Analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction