missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide (Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Description
Molecular Biology-grade Formamide is commonly used in nucleic acid hybridization and sequencing. It requires pretreatment with a mixed-bed resin.

Specifications
Specifications
pH | 4 to 5 |
Melting Point | 2°C |
Absorbance | 0.1 max. at 280nm |
Density | 1.133g/cm³ |
Boiling Point | 210°C |
Quantity | 500 mL |
Identification | Pass Test |
Packaging | Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | ≥99 % |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
- Formamide
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Formamide (Molecular Biology), Fisher BioReagents >
Spot an opportunity for improvement?Share a Content Correction