missing translation for 'onlineSavingsMsg'
Learn More
Formamide, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals Product Code.: 11400670

Formamide, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals

Product Code. 11400670
500 mL
Click to view available options
Quantity:
100 mL
500 mL
4 x 500 mL
This item is not returnable. View return policy

Product Code. 11400670

Brand: Thermo Scientific Alfa Aesar 044066.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2°C
Color Colorless
Density 1.134 g/mL
Boiling Point 210°C
Quantity 500 mL
Flash Point 154°C (310°F)
Odor Ammonia-like
Physical Form Liquid
Refractive Index 1.4475
Linear Formula HCONH2
Beilstein 505995
Merck Index 14,4237
Solubility Information Soluble in water (20mg/ml ),methanol,ethanol and acetone. Slightly soluble in ether.
Formula Weight 45.04
Percent Purity ≥99%
Grade Spectrophotometric
Chemical Name or Material Formamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453479 Certificate TypeCertificate of Analysis Date11/01/2024 Catalog Number044066.AE,044066.AY,044066.AP
Lot NumberA0442055 Certificate TypeCertificate of Analysis Date24/11/2022 Catalog Number044066.AY,044066.AP
Lot Number10243604 Certificate TypeCertificate of Analysis Date23/09/2022 Catalog Number044066.AY,044066.AP
Lot Number10240049 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044066.AY,044066.AP
Lot Number10237114 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044066.AY,044066.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formamide, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.