Learn More
Formamide, Ultra Pure, 99.5% min, Thermo Scientific Chemicals
Description
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.
Solubility
Soluble in water (20 mg/ml ), methanol, ethanol and acetone. Slightly soluble in ether.
Notes
Hygroscopic. Store at room temperature. Store away from water or moisture.

Specifications
Specifications
Melting Point | 2°C |
Color | Colorless |
Density | 1.134 g/mL |
Boiling Point | 210°C |
Quantity | 250 mL |
Flash Point | 150°C (302°F) |
Odor | Ammonia-like |
Physical Form | Liquid |
Refractive Index | 1.4475 |
Beilstein | 505995 |
Show More |
Safety and Handling

- Formamide, Ultra Pure
- Danger
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- H351-Suspected of causing cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Formamide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.