missing translation for 'onlineSavingsMsg'
Learn More

Formamide, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15548214
Click to view available options
Quantity:
100 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For denaturing nucleic acids prior to analysis

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Quantity 100 mL
Flash Point 154.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.13kg
Formula Weight 45.04
Grade Ultra Pure
Chemical Name or Material Formamide
compliance-icons
Product Identifier
  • Formamide
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Formamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'