missing translation for 'onlineSavingsMsg'
Learn More

Formamidine disulfide dihydrochloride, 97%

Product Code. 11449856
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11449856

Brand: Thermo Scientific Alfa Aesar B20331.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14807-75-1 | C2H8Cl2N4S2 | 223.13 g/mol

Formamidine disulfide dihydrochloride is used as an intermediate in agriculture and dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamidine disulfide dihydrochloride is used as an intermediate in agriculture and dyestuff intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14807-75-1
Molecular Formula C2H8Cl2N4S2
Molecular Weight (g/mol) 223.13
MDL Number MFCD00012586
InChI Key BFJQSCVWXZOXGK-UHFFFAOYSA-N
Synonym formamidine disulfide dihydrochloride, formamdine disulfide dihydrochloride, dithiourea dihydrochloride, c2h6n4s2.2hcl, carbamimidoyldisulfanylmethanimidamide dihydrochloride, formamidine disulfide dihydrochloride disulfide dihydrochloride
PubChem CID 24802135
IUPAC Name carbamimidoylsulfanyl carbamimidothioate;dihydrochloride
SMILES [Cl-].[Cl-].[NH3+]C(=N)SSC([NH3+])=N

Specifications

Melting Point ∼175°C (decomposition)
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 223.15
Percent Purity 97%
Chemical Name or Material Formamidine disulfide dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.