missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, 85%, Thermo Scientific Chemicals

Product Code. 11338406
1000 g
Click to view available options
Quantity:
1000 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Formic acid is principally used as a preservative antibacterial agent in livestock feed, in dying textiles, tanning leather and electroplating. In the poultry industry, it is sometimes added with feed to kill E. coli bacteria. It acts as a reducing agent for the catalytic reduction of nitrate in water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formic acid is principally used as a preservative antibacterial agent in livestock feed, in dying textiles, tanning leather and electroplating. In the poultry industry, it is sometimes added with feed to kill E. coli bacteria. It acts as a reducing agent for the catalytic reduction of nitrate in water.

Solubility
Miscible with water, ether, acetone, ethyl acetate, glycerol, methanol and ethanol. Partially miscible with benzene, toluene and xylene.

Notes
Hygroscopic. Store closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Melting Point 7°C to 9°C
Boiling Point 100°C to 101°C
Linear Formula HCOOH
UN Number UN1779
Beilstein 1209246
Merck Index 14,4241
Solubility Information Miscible with water,ether,acetone,ethyl acetate,glycerol,methanol and ethanol. Partially miscible with benzene,toluene and xylene.
Formula Weight 46.03
Percent Purity 85%
Density 1.22
Quantity 1000 g
Flash Point 50°C (156°F)
Refractive Index 1.371
Sensitivity Hygroscopic
Chemical Name or Material Formic acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid, 85%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P372-Explosion risk in case of fire.
  • P374-Fight fire with normal precautions from a reasonable distance.
  • P380-Evacuate area.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P404-Store in a closed container.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : formic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formic acid, 85%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.