Learn More
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
32.40 € - 98.40 €
Chemical Identifiers
CAS | 64-18-6 |
---|---|
Molecular Formula | CH2O2 |
Molecular Weight (g/mol) | 46.025 |
MDL Number | MFCD00003297 |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
Synonym | methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt |
PubChem CID | 284 |
ChEBI | CHEBI:30751 |
IUPAC Name | formic acid |
SMILES | C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10259750
|
Thermo Scientific Acros
423755000 |
500 g | Plastic bottle |
32.40 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10628914
|
Thermo Scientific Acros
423750025 |
2.5 kg | Plastic bottle |
98.40 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-18-6 | |
46.025 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
CH2O2 | |
MFCD00003297 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
CHEBI:30751 | |
C(=O)O |
Specifications
64-18-6 | |
101.0°C | |
CH2O2 | |
MFCD00003297 | |
01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.02 | |
ACS Reagent | |
50°C | |
Liquid | |
0.003% max. | |
44mbar at 20°C |
8.0°C | |
97% | |
HCO2H | |
02, 8 | |
15, 4269 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol, ether and glycerine | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
97% | |
1.2200g/mL | |
Authentic | |
1.3700 to 1.3720 | |
1.22 | |
Formic acid, 96+% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 200-579-1
missing translation for 'rtecsNumber' : LQ4900000
missing translation for 'tsca' : TSCA
- Formic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only