missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 98-100%, Extra Pure, SLR, Fisher Chemical™

32.25 € - 107.00 €

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number 3297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10375990
View Documents
Fisher Chemical
F/1850/PB08
500 mL
32.25 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10478020
View Documents
Fisher Chemical
F/1850/PB17
2.5 L
107.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

64-18-6
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
284
formic acid
CH2O2
3297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
CHEBI:30751
C(=O)O

Specifications

2.1
8°C
Colorless
3297
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
284
46.02g/mol
Liquid
64-18-6
101°C
CH2O2
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
C(=O)O
46.025
CHEBI:30751
Plastic Coated Glass bottle
44mbar at 20°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Formic acid 98/100%

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications