missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic acid, 98+%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Assay Percent Range | 98+% |
Linear Formula | HCO2H |
Beilstein | 02, 8 |
Fieser | 01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177 |
Merck Index | 15, 4269 |
Formula Weight | 46.02 |
Percent Purity | ≥98.0% |
Grade | Pure |
Density | 1.2200g/mL |
Quantity | 25 mL |
Show More |
Safety and Handling
Product Identifier
- Formic acid
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- EUH071-Corrosive to the respiratory tract.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction