missing translation for 'onlineSavingsMsg'
Learn More
Formic acid, ACS, 96+%, Thermo Scientific Chemicals Product Code.: 11358058

Formic acid, ACS, 96+%, Thermo Scientific Chemicals

Product Code. 11358058
2 kg
Click to view available options
Quantity:
500 g
2 kg
4 x 500 g
This item is not returnable. View return policy

Product Code. 11358058

Brand: Thermo Scientific Alfa Aesar 036617.A3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a hydrogen source, as a reducing agent and is used in tanning and dye fixing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Melting Point 7°C to 9°C
Boiling Point 100°C to 101°C
Color Colorless
Assay Percent Range >96+%
Linear Formula HCOOH
UN Number UN1779
Beilstein 1209246
Merck Index 14,4241
Solubility Information Miscible with water. Partially miscible in toluene,xylene and benzene.
Formula Weight 46.03
Percent Purity ≥96%
Grade ACS
Density 1.220 g/mL
Quantity 2 kg
Flash Point 50°C (156°F)
Physical Form Liquid
Refractive Index 1.371
Sensitivity Hygroscopic
Chemical Name or Material Formic acid, For analysis ACS
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP14L032 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog Number036617.57,036617.A3,036617.36
Lot NumberM28L044 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number036617.57,036617.A3,036617.36
Lot NumberW16K024 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number036617.57,036617.A3,036617.36
Lot NumberS27K026 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number036617.57,036617.A3,036617.36
Lot NumberZ20J046 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog Number036617.57,036617.A3,036617.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formic acid, ACS, 96+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.